Would you like to log in again?
Human Serpin C1 - Antithrombin-III ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0839
76236-234EA 636 USD InStock
76236-234
Human Serpin C1 - Antithrombin-III ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Serpin C1 - Antithrombin-III AccuSignal ELISA Kit
  • Natural and recombinant human Serpin C1.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: H33-K464
Antithrombin III is the most important inhibitor of thrombin and other coagulation proteinases. It belongs to the serine proteinase inhibitor (serpin) superfamily of inhibitors and structurally related proteins, which contain reactive centers that have evolved to attract and entrap certain proteinases. Inherited antithrombin III deficiency (AT3D) is a risk factor for the early development of venous thromboembolism (THPH7). Antithrombin III regulates clot formation both by inhibiting thrombin activity directly and by interfering with earlier stages of the clotting cascade. By in situ hybridization and quantitative analysis of DNA dosage in carriers of chromosome 1 deletions, AT3 is mapped to 1q23-q25. The AT3 gene has 7 exons. It contains 9 complete and 1 partial repetitive ALU sequence elements, which occur in the introns of the gene at a higher frequency (about 22% of the intron sequence) than in the genome as a whole (about 5%).
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 20,000pg/ml, 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml human Serpin C1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma (heparin, EDTA, citrate) or urine to each empty well. We recommend that each human Serpin C1 standard solution and each sample is measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty