Would you like to log in again?
Human CD5L ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0853
76236-238EA 636 USD InStock
76236-238
Human CD5L ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human CD5L AccuSignal ELISA Kit
  • Natural and recombinant human CD5L.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: S20-G347
CD5 antigen-like, also known as Sp alpha and AIM, is a protein that in humans is encoded by the CD5L gene. It is mapped to 1q21-q23 by fluorescence in situ hybridization. It is found that Aim expression is induced in mouse macrophages in response to loading with highly oxidized low density lipoprotein (oxLDL), and that Aim is expressed in foam cells within atherosclerotic lesions. Both the expression of Aim in lesions and its induction by oxLDL require Lxr /Rxr heterodimers. Aim-null macrophages are highly susceptible to oxLDL-induced apoptosis in vitro and undergo accelerated apoptosis in atherosclerotic lesions in vivo. Double knockout of Aim and Ldlr reduce atherosclerotic lesions. Therefore, it is concluded that AIM expression protects macrophages from apoptosis within atherosclerotic lesions, promoting early lesion development.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 313pg/ml, 156pg/ml human CD5L standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma (heparin, EDTA) to each empty well. It is recommended that each human CD5L standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty