Would you like to log in again?
Human AGRP - agouti-related ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0862
76236-256EA 636 USD InStock
76236-256
Human AGRP - agouti-related ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human AGRP - agouti-related AccuSignal ELISA Kit
  • Natural and recombinant human AGRP.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: sf21; Immunogen sequence: A21-T132
Agouti-related protein, also called Agouti-related peptide (AgRP), is a neuropeptide produced in the brain by the AgRP/NPY neuron. It is only synthesised in NPY-containing cell bodies located in the ventromedial part of the arcuate nucleus in the hypothalamus. In humans, the agouti-related peptide is encoded by the AGRP gene. AgRP is co-expressed with Neuropeptide Y and works by increasing appetite and decreasing metabolism and energy expenditure. It is one of the most potent and long-lasting of appetite stimulators.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.3pg/ml, 15.6pg/ml, 7.8pg/ml human AGRP standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma(heparin, EDTA, citrate) or urine to each empty well. It is recommended that each human AGRP standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty