Would you like to log in again?
AccuSignal Genomic DNA Magnetic Bead Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
KOA0912
76236-386EA 533 USD InStock
76236-386
AccuSignal Genomic DNA Magnetic Bead Kit, Rockland Immunochemicals
Assays Immunological Assays
AccuSignal Genomic DNA Magnetic Bead Kit
  • extraction and isolation of genomic DNA
  • Bead mean diameter is ~0.5 μm, Bead concentration is 12.5 mg/mL
  • Binding capacity varies with sample type
  • contains magnetic beads and reagents sufficient for 100 samples
The Trueblot® Genomic DNA Magnetic Beads Kit is designed to purify genomic DNA from mammalian tissues and bacteria. Paramagnetic beads with uniform particle size efficiently bind DNA, resulting in high yields of DNA with minimal RNA, proteins, nucleases, and other cellular contaminants. The kit is intended for manual purifications using a magnetic separator. The protocol can be easily customized to optimize sample yield and quality depending on the type of tissue or bacteria. The kit contains magnetic beads and reagents sufficient for 100 samples. Magnetic separator is available separately. The uniform size and surface area of beads ensure highly reproducible results and improved performance compared to magnetic beads from alternative suppliers. Bead mean diameter is ~0.5 μm, Bead concentration is 12.5 mg/mL and Binding capacity varies with sample type.
The Trueblot® Genomic DNA Magnetic Beads Kit can be used for extraction and isolation of genomic DNA from bacteria and mammalian cells and tissue samples.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty