Would you like to log in again?
Tris-Acetate-EDTA (TAE) 50X, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
MB-020-4000
76236-398EA 368 USD InStock
76236-398
Tris-Acetate-EDTA (TAE) 50X, Rockland Immunochemicals
Buffers Electrophoresis Buffers Running Buffers
Tris-Acetate-EDTA (TAE) is a buffer solution containing a mixture of Tris base, acetic acid and EDTA.
  • Concentrated 50X
  • 2.0 M Tris-Acetate, 0.05 M EDTA at a pH of 8.3
  • Molecular biology applications including agarose electrophoresis for the separation of nucleic acids such as DNA and RNA
TAE is suitable for multiple molecular biology applications including agarose electrophoresis for the separation of nucleic acids such as DNA and RNA.
Application Note: Tris-Acetate-EDTA (TAE) is a concentrated stock solution and should be diluted appropriately with distilled, deionized water or equivalent to its final working concentration. Tris-Acetate-EDTA (TAE) consists of 2.0 M Tris-Acetate, 0.05 M EDTA at a pH of 8.3.  Meticulously prepared using ultra pure reagents dissolved in highly polished pharmaceutical grade deionized water.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty