Would you like to log in again?
Tris HCl pH 8.5, 1.0 M
Supplier: Rockland Immunochemicals
MB-027-1000
76236-402EA 82.82 USD InStock
76236-402
Tris HCl pH 8.5, 1.0 M
Buffers Biological Buffers
Tris solution is an abbreviation of the organic compound known as tris(hydroxymethyl)aminomethane. Tris is extensively used in biochemistry and molecular biology.
  • This product is a concentrated stock solution and should be diluted appropriately with distilled, deionized water or equivalent to its final working concentration.
  • This buffer consists of 1.0 M Tris HCl at pH 8.5.
In biochemistry, Tris is widely used as a component of buffer solutions, such as in TAE and TBE buffer, especially for solutions of nucleic acids.
Application Note: Tris solution is suitable for molecular immunology, molecular biology and nucleic acid methodologies, when applicable. Visit our newly expanded web site at www.rockland-inc.com for methods using this and other buffers. This product is a concentrated stock solution and should be diluted appropriately with distilled, deionized water or equivalent to its final working concentration. This buffer consists of 1.0 M Tris HCl at pH 8.5.  Meticulously prepared using ultra pure reagents dissolved in highly polished pharmaceutical grade (WFI) water.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty