Would you like to log in again?
10X Tris-Glycine
Supplier: Rockland Immunochemicals
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
MB-029-1000 MB-029-4000
76236-406EA 94 USD InStock
76236-406 76236-408
10X Tris-Glycine
Buffers Electrophoresis Buffers Running Buffers
Tris-Glycine Running Gel buffer for polyacrylamide gel electrophoresis - without SDS (sodium dodecyl sulfate)
  • Concentrated 10X
  • Tris-Glycine Running Gel buffer for polyacrylamide gel electrophoresis
  • Without SDS (sodium dodecyl sulfate)
Tris-Glycine Running Gel buffer without SDS (sodium dodecyl sulfate) for polyacrylamide gel electrophoresis, describes a technique widely used in biochemistry to separate proteins according to their electrophoretic mobility (a function of the length of a polypeptide chain and its charge) and no other physical feature. SDS is an anionic detergent applied to protein sample to linearize proteins and to impart a negative charge to linearized proteins.
Application Note: Tris-Glycine running buffer is suitable for laboratory involved in protein biochemistry. Visit the newly expanded web site at www.rockland-inc.com for methods using this and other buffers. This product is a 10X concentrated stock solution and should be diluted appropriately with distilled, deionized water (or equivalent) to its final working concentration. No pH adjustment is required. 10X Tris-Glycine Running Buffer consists of 0.2 M Tris HCl, 1.5 M Glycine); pH 8.0.  Meticulously prepared using ultra-pure reagents dissolved in highly polished pharmaceutical grade deionized water.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty