Would you like to log in again?
10X Tris-Acetate-EDTA (TAE)
Supplier: Rockland Immunochemicals
MB-039-1000
76236-410EA 112 USD InStock
76236-410
10X Tris-Acetate-EDTA (TAE)
Buffers Electrophoresis Buffers Running Buffers
10X Tris-Acetate-EDTA (TAE) Buffer
  • Concentrated 10X
  • 0.4 M Tris-Acetate, 0.01 M EDTA at a pH of 8.5
  • Concentrated stock solution and should be diluted appropriately with distilled, deionized water or equivalent to its final working concentration
Tris-Acetate-EDTA (TAE) is a buffer solution containing a mixture of Tris base, acetic acid and EDTA. TAE suitable for multiple molecular biology applications including agarose electrophoresis for the separation of nucleic acids such as DNA and RNA.
Application Note: Tris-Acetate-EDTA (TAE) is a concentrated stock solution and should be diluted appropriately with distilled, deionized water or equivalent to its final working concentration. Tris-Acetate-EDTA (TAE) consists of 0.4 M Tris-Acetate, 0.01 M EDTA at a pH of 8.5.  Meticulously prepared using ultra pure reagents dissolved in highly polished pharmaceutical grade deionized water.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty