Would you like to log in again?
Sodium Dodecyl Sulfate SDS, 20%
Supplier: Rockland Immunochemical
MB-046-1000 MB-046-0100
76236-416EA 112 USD InStock
76236-416 76236-418
Sodium Dodecyl Sulfate SDS, 20%
Buffers Extraction Buffers
SDS, sodium dodecyl sulfate is an anionic surfactant.
  • 20% (w/v) Sodium Dodecyl Sulfate (SDS)
  • Aseptically filtered through a Millipore 0.22 micron filter into clean, pre-sterilized containers
  • Tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria
SDS is commonly used in laboratory as component of buffer for cell lysis, cell lysis during DNA extraction and mostly in SDS-PAGE running buffer. Indeed, SDS is an anionic detergent applied to protein sample to linearize proteins and to impart a negative charge to linearized proteins.
Application Note: This product is a concentrated stock solution and should be diluted appropriately with distilled, deionized water or equivalent to its final working concentration. Meticulously prepared using ultra pure reagents dissolved in highly polished pharmaceutical grade deionized water.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty