Would you like to log in again?
1X PBS Buffer with 1% Casein
Supplier: Rockland Immunochemicals
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
MB-081-0100 MB-081-1000
76236-542EA 79 USD InStock
76236-542 76236-544
1X PBS Buffer with 1% Casein
Buffers Electrophoresis Buffers Washing Buffers
Phosphate buffered saline with casein is suitable for multiple applications including biological diluent buffer for antibodies or other biologics.
  • Ready-to-Use
  • Diluent or wash buffer for assays
  • Aseptically filtered through a Millipore 0.22 micron filter into clean, pre-sterilized containers
  • Tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria
It also may be used as a wash buffer for immunological assays including western blot, immunohistochemistry, immunofluorescence microscopy, and ELISA. Other applications may require detergents or other additional components.
Application Note: This product is a ready-to-use 1X solution and no further dilution is required.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty