Would you like to log in again?
Mouse IGFBP-1 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0200
76236-624EA 652 USD InStock
76236-624
Mouse IGFBP-1 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse IGFBP-1 AccuSignal ELISA Kit
  • Natural and recombinant mouse IGFBP-1.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: A26-N272
IGFBP1 is also known as amniotic fluid binding protein(AFBP), placental protein-12, alpha-pregnancy-associated endometrial globulin, growth hormone independent binding protein, binding protein-28, binding protein-26, and binding protein-25. The low-molecular weight insulin-like growth-factor binding protein(IGF-BP25) is synthesized by human liver, secretory endometrium and decidua, and is also present in human serum. It binds insulin-like growth factors IGF-I and IGF-II with high affinity, and is proposed to act as a paracrine regulator of cell growth. These IGF-binding proteins are expressed at different concentrations in different tissues and are thought to regulate the activity of IGF I and II. The gene is organized in four exons and spans 5.9 kb. The IBP-1 gene is a single copy gene, located on chromosome 7. The standard product used in this kit is recombinant mouse IGFBP-1, consisting of 247 amino acids with the molecular mass of 27KDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml mouse IGFBP-1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse IGFBP-1 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty