Would you like to log in again?
Mouse IGFBP-3 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0204
76236-632EA 652 USD InStock
76236-632
Mouse IGFBP-3 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse IGFBP-3 AccuSignal ELISA Kit
  • Natural and recombinant mouse IGFBP-3.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: G28Q-K291
Insulin-like growth factor(IGF)-binding protein-3(IGFBP-3) is a major determinant of circulating levels of the IGFs and is clinically useful for the evaluation of GH deficiency and for predicting the response to GH treatment. The circulating level of IGFBP-3 is inversely related to the risk of several common cancers, and that antiproliferative agents such as antiestrogens and retinoids act in part by up-regulating IGFBP-3 gene(IGFBP3) expression. Insulin-like growth factor-binding protein(IGFBP)-3, well characterized as the carrier of insulin-like growth factor(IGF), has been reported to have intrinsic bioactivity that is independent of IGF binding. IGFBP-3 has an IGF-independent, antiproliferative effect in undifferentiated and early differentiated but not in terminally differentiated chondrocytes. IGFBP-3 possesses both growth-inhibitory and potentiating effects on cells that are independent of IGF action and are mediated through specific. IGFBP-3 binding proteins/receptors locate at the cell membrane, cytosol, or nuclear compartments and in the extrIGFBP-3llular matrix. IGFBP3 is also located on chromosome 7. The standard product used in this kit is recombinant mouse IGFBP-3, consisting of 270 amino acids with the molecular mass of 29.4kDa. As a result of glycosylation, the molecular mass is 40-50KDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 313pg/ml, 156pg/ml mouse IGFBP-3 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates or serum to each empty well. It is recommended that each mouse IGFBP-3 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty