Would you like to log in again?
Human IL-1 alpha ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0206
76236-636EA 618 USD InStock
76236-636
Human IL-1 alpha ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Interleukin-1 alpha AccuSignal ELISA Kit
  • Natural and recombinant human IL-1 alpha.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: S113-A271
Interleukin-1 alpha(IL-1 alpha) and interleukin-1 beta(IL-1 beta) are two biochemically distinct, but distantly related, polypeptidic cytokines that play a key role in inflammation, immunologic reactions, and tissue repair. IL-1 alpha has been implicated in the pathogenesis of infectious, autoimmune and inflammatory diseases. Recently, it has been shown that IL-1 alpha is identical to hematopoietin 1, which is described as a hematopoietic growth factor acting on early progenitor cells in synergy with other hematopoietic growth factors. The human interleukin 1 alpha gene is assigned to chromosome 2. Genetic polymorphisms at interleukin(IL)-1alpha and IL-1beta have been recently suggested to be associated with severity of adult periodontitis. The murine IL-1 alpha and IL-1 beta genes encode structurally and evolutionarily related cytokines that exert a regulatory role in numerous physiological processes including hemopoiesis. The standard product used in this kit is recombinant human IL-1alpha with the molecular mass of 18kDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 300pg/ml, 150pg/ml, 75pg/ml, 37.5pg/ml, 18.8pg/ml, 9.4pg/ml, 4.7pg/ml human IL-1 alpha standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA, citrate) to each empty well. We recommend that each human IL-1 alpha standard solution and each sample is measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty