Would you like to log in again?
Human IL1R2 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0213
76236-650EA 642 USD InStock
76236-650
Human IL1R2 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human IL1R2 AccuSignal ELISA Kit
  • Natural and recombinant human IL1R2.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: sf21; Immunogen sequence: F14-E343
Interleukin 1 receptor, type II(IL1R2) also known as CD121b(Cluster of Differentiation 121b) is an interleukin receptor. The protein encoded by this gene is a cytokine receptor that belongs to the interleukin-1 receptor family. It is mapped to 2q11.2. IL1R2 binds interleukin alpha(IL1A), interleukin beta(IL1B), and interleukin 1 receptor, type I(IL1R1/IL1RA), and acts as a decoy receptor that inhibits the activity of its ligands. IL1R2 also modulates cellular response through non-signaling association with IL1RAP after binding to IL1B. The secreted IL1R2 recruits secreted IL1RAP with high affinity, and this complex formation may be the dominant mechanism for neutralization of IL1B by secreted/soluble receptors.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml human IL1R2 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates or serum to each empty well. It is recommended that each human IL1R2 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty