Would you like to log in again?
Human IL-5 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0223
76236-670EA 636 USD InStock
76236-670
Human IL-5 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Interleukin-5 AccuSignal ELISA Kit
  • Natural and recombinant human IL-5.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: SF21; Immunogen sequence: I20-S134
Interleukin-5(IL-5) is also known as eosinophil differentiation factor(EDF). IL-5 is a potential candidate gene in the pathogenesis of asthma, as it is the main cytokine controlling eosinophil activity and eosinophils are pivotal in the development of airway inflammation. The predicted amino acid sequence of 134 amino acids is identical with that recently reported for human interleukin-5 but shows no significant homology with other known hemopoietic growth regulators. IL-5 is a lineage-specific hematopoietic growth factor that stimulates the production of eosinophils and eosinophil colonies from normal human bone marrow cells. IL-5 gene to human chromosome 5 at bands q23-31. Human and mouse of IL-5 share 70% amino acid sequence homology.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.3pg/ml, 15.6pg/ml, 7.8pg/ml human IL-5 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma(heparin, EDTA) or urine to each empty well. It is recommended that each human IL-5 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty