Would you like to log in again?
Mouse IL-6 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
KOA0226
76236-676EA 636 USD InStock
76236-676
Mouse IL-6 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse Interleukin-6 AccuSignal ELISA Kit
  • Natural and recombinant mouse IL-6.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: F25-T211
The human interferon-beta 2 gene(IFNB2) product is identical to that for the B-cell stimulation factor-2(BSF-2), the hybridoma growth factor(HGF)("interleukin-6"), and the hepatocyte stimulating factor(HSF). Proteins derived from this gene mediate the plasma protein response to tissue injury(acute-phase response) and regulate the growth and differentiation of both B and T cells. Interleukin-6(IL6) has come to be regarded as a potential osteoporotic factor because it has stimulatory effects on cells of the osteoclast lineage, and, thus, may play a role in the pathogenesis of bone loss associated with estrogen deficiency. IL-6 has many roles essential to the regulation of the immune response, hematopoiesis, and bone resorption. It is involved not only in the hepatic acute phase response but also in adipose tissue metabolism, lipoprotein lipase activity, and hepatic triglyceride secretion. Overproduction of IL-6, a proinflammatory cytokine, is associated with a spectrum of age-related conditions including cardiovascular disease, osteoporosis, arthritis, type 2 diabetes, certain cancers, periodontal disease, frailty, and functional decline. BSF-2 is a novel interleukin consisting of 184 amino acids. The standard product used in this kit is recombinant IL-6, consisting of 188 amino acids with the molecular mass of 21.8kDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml mouse IL-6 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA, citrate) to each empty well. It is recommended that each mouse IL-6 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty