Would you like to log in again?
Mouse IL-10 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0230
76236-684EA 636 USD InStock
76236-684
Mouse IL-10 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse Interleukin-10 AccuSignal ELISA Kit
  • Natural and recombinant mouse IL-10.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: S19-S178
Interleukin-10, also called cytokine synthesis inhibitory factor, is implicated in tumorigenesis, and it has been shown that polymorphisms in its gene promoter correlate with differential amounts of production. IL-10 is an important cytokine with anti-inflammatory, anti-immune, and antifibrotic functions. It is also an important regulatory cytokine whose involvement extends into diverse areas of the human immune system. IL-10 is a recently described natural endogenous immunosuppressive cytokine that has been identified in human, murine, and other organisms. IL-10 significantly affects chemokine biology, because human IL-10 inhibits chemokine production and is a specific chemotactic factor for CD8+ T cells. It suppresses the ability of CD4+ T cells, but not CD8+ T cells, to migrate in response to IL-8. Interleukin-10 gene polymorphisms and interleukin-10 production capability may contribute to the development of skin squamous cell carcinomas after renal transplantation. The interleukin-10 locus contributes to the heritability of psoriasis susceptibility. With regard to sudden infant death, IL-10 is of special interest. This is an immunoregulatory cytokine that plays an important role in the development of infectious disease. The mIL-10 gene is mapped to mouse chromosome 1 and the hIL-10 gene is also mapped to human chromosome 1. The standard product used in this kit is recombinant mouse IL-10, consisting of 160 amino acid with the molecular mass of 18KDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml mouse IL-10 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates or serum to each empty well. It is recommended that each mouse IL-10 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty