Would you like to log in again?
Mouse IL-13 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0238
76236-692EA 724 USD InStock
76236-692
Mouse IL-13 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse Interleukin-13 AccuSignal ELISA Kit
  • Natural and recombinant mouse IL-13.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: S26-F131
Interleukin(IL)-13 is a major inducer of fibrosis in many chronic infectious and autoimmune diseases. Recombinant IL-13 protein inhibits inflammatory cytokine production induced by lipopolysaccharide in human peripheral blood monocytes. Moreover, it synergizes with IL-13 in regulating interferon-gamma synthesis in large granular lymphocytes. Interleukin-13 may be critical in regulating inflammatory and immune responses. IL-13 and IL-4 are two cytokines produced by T helper type 2 cells, mast cells, and basophils. In addition to their physiological roles, these cytokines are also implicated in pathological conditions such as asthma and allergy. The genes encoding human IL-4 and IL-13 are located on segment q23-31 of chromosome 5.4 Recombinant mouse IL-13 is a mixture of Ser26-Phe131 amino acids with the molecular mass of 11.5KDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml mouse IL-13 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse IL-13 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty