Would you like to log in again?
Mouse Leptin receptor ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0249
76236-710EA 652 USD InStock
76236-710
Mouse Leptin receptor ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse Leptin receptor AccuSignal ELISA Kit
  • Natural and recombinant mouse Leptin receptor.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence:  L22-G839
Leptin receptor(or Obese receptor, OBR) is a single membrane-spanning receptor most related to the gp130 signal-transducing component of the IL-6 receptor, the G-CSF receptor, and the LIF receptor. OB-R mRNA is expressed not only in choroid plexus, but also in several other tissues, including hypothalamus. Leptin acts through the leptin receptor, a single-transmembrane domain receptor of the cytokine-receptor family. Leptin controls energy balance and food intake through the leptin receptor in the hypothalamus of the brain, which suggests that some polymorphisms of the leptin receptor gene(LEPR) might contribute to obesity or obesity-related diseases. Leptin is also involved in the regulation of blood pressure through the leptin receptor.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml mouse Leptin receptor standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse Leptin receptor standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty