Would you like to log in again?
Human MMP-3 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0269
76236-742EA 652 USD InStock
76236-742
Human MMP-3 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human MMP-3 AccuSignal ELISA Kit
  • Natural and recombinant human total MMP-3.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence:   Y18-C477
Matrix metalloproteinase-3(MMP-3) also called stromelysin or transin, is a proteoglycanase closely related to collagenase(MMP1) with a wide range of substrate specificities. The complete primary structure for human MMP-3, which has 477 residues including a 17-residue signal peptide. MMP-3 and collagenase are 54% identical in sequence, suggesting a common origin for the evolution of the two proteinases. MMP-3 and collagenase expression are coordinately modulated in synovial fibroblast cultures. MMP-3 is a secreted metalloprotease produced predominantly by connective tissue cells. Together with other metalloproteases, it can synergistically degrade the major components of the extracellular matrix. It is capable of degrading proteoglycan, fibronectin, laminin, and type IV collagen, but not interstitial type I collagen. MMP-3 genotype may be an important determinant of vascular remodeling and age-related arterial stiffening, with the heterozygote having the optimal balance between matrix accumulation and deposition. The standard product used in this kit is recombinant human MMP-3, consisting of 460 amino acids with the molecular mass of 52KDa. The detected MMP-3 includes zymogen and active enzyme.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human MMP-3 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma( heparin) to each empty well. It is recommended that each human MMP-3 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty