Would you like to log in again?
Human MMP-7 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0271
76236-746EA 724 USD InStock
76236-746
Human MMP-7 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human MMP-7 AccuSignal ELISA Kit
  • Natural and recombinant human total MMP-7.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence:  L18-K267
Matrix metalloproteinase-7(MMP-7) previously called putative metalloproteinase I(PUMP1) or matrilysin. The PUMP1 gene has been identified through studies of collagenase-related connective-tissue-degrading metalloproteinases produced by human tumors. The PUMP I protein has 267 amino acids and is significantly shorter than stromelysin or collagenase(477 and 469 amino acids, respectively). Matrix metalloproteinases play a crucial role in tumor invasion and metastasis. Matrilysin, a member of the matrix metalloproteinase family, is structurally different from the other matrix metalloproteinases by virtue of the absence of a conserved COOH-terminal protein domain. In addition, matrilysin mRNA is regulated in a specific and distinct manner in normal and malignant tissues. Matrilysin has been shown to correlate with nodal or distant metastasis in colorectal carcinomas; however, its implication in early invasive colorectal carcinomas has not been determined.1 Matrilysin is also a mediator of pulmonary fibrosis and a ...
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human MMP-7 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma(heparin), saliva or urine to each empty well. It is recommended that each human MMP-7 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty