Would you like to log in again?
Human MMP-9 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0273
76236-750EA 724 USD InStock
76236-750
Human MMP-9 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human MMP-9 AccuSignal ELISA Kit
  • Natural and recombinant human total MMP-9.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence:    A20-D707
The 92-kD type IV collagenase is also known as 92-kD gelatinase, type V collagenase, gelatinase B, or matrix metalloproteinase-9(MMP9). The 72- and 92-kDa type IV collagenases are members of a group of secreted zinc metalloproteases. The matrix metalloproteinases(MMPs) are able to degrade the extracellular matrix and allow angiogenesis and tumor invasion. Gelatinase B, a matrix metalloproteinase that has proteolytic activity against connective tissue proteins, has been suggested to be important in the connective tissue remodeling processes associated with atherogenesis and plaque rupture. MMP-9 is predominantly expressed in neutrophils, macrophages, and mast cells, rather than in oncogene-positive neoplastic cells. The polymorphism of MMP-9 acts as a genetic factor for the development of smoking-induced pulmonary emphysema. The standard product used in this kit is recombinant human MMP-9 with the molecular mass of 95KDa. The detected MMP-9 includes zymogen and active enzyme.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human MMP-9 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma(heparin), saliva or urine to each empty well. It is recommended that each human MMP-9 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty