Would you like to log in again?
Rat PDGF-AB ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0290
76236-776EA 636 USD InStock
76236-776
Rat PDGF-AB ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Rat PDGF-AB AccuSignal ELISA Kit
  • Natural and recombinant rat PDGF-AB.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: (R86-K204)+(S74-T182)
The platelet-derived growth factor(PDGF) is a mitogen derived from human platelets consisting of two related polypeptides termed A and B chains. The genes for PDGF A chain, B chain/c-sis, and the PDGF receptor are expressed in human malignant glioma cell lines. Normal human endothelial cells in culture express the B chain of PDGF, and that endothelial-derived PDGF B chain is synthesized as a predicted precursor polypeptide of Mr 27,281. The entire B chain of PDGF is highly(96%) homologous to a portion of p28sis, the transforming protein of simian sarcoma virus(SSV). It has been suggested that p28sis exerts its transforming potential by mimicking the growth promoting activity of PDGF and stimulating the cell in an autocrine manner. PDGF A-chain precursor polypeptide is assigned to the proximal long arm of chromosome 7, band q11.23. The human homolog(PDGF B-chain/c-sis) of the transforming gene of simian sarcoma virus is assigned to chromosome 22. The standard product used in this kit is recombinant rat PDGF-AB...
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml rat PDGF-AB standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of rat cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each rat PDGF-AB standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty