Would you like to log in again?
Human TIMP-3 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0315
76236-792EA 687 USD InStock
76236-792
Human TIMP-3 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human TIMP-3 AccuSignal ELISA Kit
  • Natural and recombinant human TIMP-3.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: C24-P211
The tissue inhibitors of metalloproteinases(TIMPs) are natural inhibitors of the matrix metalloproteinases, a group of zinc-binding endopeptidases involved in the degradation of the extracellular matrix. The TIMP3 gene is expressed in many tissues, with highest expression in the placenta. TIMP3 encodes a potent angiogenesis inhibitor and is mutated in Sorsby fundus dystrophy, a macular degenerative disease with submacular choroidal neovascularization. TIMP3 gene is mapped to 22q12.1-q13.2. Mutations in TIMP3 cause the autosomal dominant disorder Sorsby's fundus dystrophy(SFD).
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human TIMP-3 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma(heparin , EDTA) or saliva to each empty well. It is recommended that each human TIMP-3 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty