Would you like to log in again?
Human sTNFsR II ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0322
76236-806EA 636 USD InStock
76236-806
Human sTNFsR II ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human sTNFsR II AccuSignal ELISA Kit
  • Natural and recombinant human sTNFsRⅡ.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: L23-D257
Tumor necrosis factor receptor p75(TNF-R p75, or TNF-R2) is a 75-kDa type I transmembrane protein expressed predominantly on cells of hematopoietic lineage. TNF-R p75 belongs to the TNF receptor superfamily characterized by cysteine-rich extracellular regions composed of three to six disulfide-linked domains. The tumor necrosis factor receptor II(TNFRII) gene localizes to 1p36. 2, a genomic region characteristically deleted in neuroblastomas and other malignancies. In addition, TNFRII is the principal mediator of the effects of TNF on cellular immunity, and it may cooperate with TNFRI in the killing of nonlymphoid cells. The standard product used in this kit is recombinant human sTNFRII, consisting of 24-206 amino acid sequence with the molecular mass of 20KDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.3pg/ml, 15.6pg/ml, 7.8pg/ml human sTNFsRⅡ standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma( heparin, EDTA, citrate) or urine to each empty well. We recommend that each human sTNFsRⅡ standard solution and each sample is measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty