Would you like to log in again?
Human uPA - PLAU ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0325
76236-812EA 636 USD InStock
76236-812
Human uPA - PLAU ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human uPA - PLAU AccuSignal ELISA Kit
  • Natural and recombinant human Upa.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: M1-L431
Plasminogen activator,urokinase(PLAU, uPA) converts plasminogen to plasmin. Plasmin is involved in processing of amyloid precursor protein and degrades secreted and aggregated amyloid-beta, a hallmark of Alzheimer disease(AD). Urokinase has a molecular mass of about 54 kD and is composed of 2 disulfide-linked chains, A and B, of molecular masses 18 kD and 33 kD, respectively. It localized on 10q24. uPA facilitates cell migration by localizing proteolisys on the cell surface and by inducing intracellular signalling pathways. In human vascular smooth muscle cell(VSMC), uPA stimulates migration via the uPA receptor(uPAR) signalling complex containing TYK2 and phosphatidylinositol 3-kinase(PI3-K).
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml human uPA standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human uPA standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty