Would you like to log in again?
Mouse VEGF ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0331
76236-824EA 636 USD InStock
76236-824
Mouse VEGF ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse VEGF AccuSignal ELISA Kit
  • This kit recognizes both the 164 and 120 amino acid residue forms of mouse VEGF.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: sf21; Immunogen sequence: A27-R190
Vascular permeability factor/vascular endothelial growth factor(VPF/VEGF), a potent cytokine expressed by most malignant tumors, has critical roles in vasculogenesis and both physiological and pathological angiogenesis. VEGF produced by tumor cells potently stimulates endothelial cell proliferation and angiogenesis and plays a key role in the pathophysiology of several neoplasias. VEGF may also play a pivotal role in mediating the development and progression of diabetic retinopathy. VEGF, a major regulator of angiogenesis, binds to two receptor tyrosine kinases, KDR/Flk-1 and Flt-1.The VEGF gene is mapped by fluorescence in situ hybridization to chromosome 6p12. The standard product used in this kit is recombinant mouse VEGF164, a 42KDa dimer.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.3pg/ml, 15.6pg/ml mouse VEGF standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA, citrate) to each empty well. We recommend that each mouse VEGF standard solution and each sample is measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty