Would you like to log in again?
Human VEGFR2 - KDR ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0333
76236-828EA 687 USD InStock
76236-828
Human VEGFR2 - KDR ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human VEGFR2 - KDR AccuSignal ELISA Kit
  • Natural and recombinant human VEGFR2.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: A20-E764
Vascular endothelial growth factor receptor 2(VEGFR-2) also known as Kinase insert domain receptor(KDR, a type III receptor tyrosine kinase) is a VEGF receptor. Through in situ hybridization of a genomic DNA probe to metaphase chromosomes, VEGFR2 was localized to 4q11-->q12. VEGF receptor 2 and the adherens junction act as shear-stress cotransducers, mediating the transduction of shear-stress signals into vascular endothelial cells.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml human VEGFR2 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. We recommend that each human VEGFR2 standard solution and each sample is measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty