Would you like to log in again?
Human ACE ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
KOA0337
76236-836EA 754 USD InStock
76236-836
Human ACE ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human ACE AccuSignal ELISA Kit
  • Natural and recombinant human ACE.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: sf2NSO; Immunogen sequence: L30-L1261
Angiotensin-converting enzyme(ACE) is a zinc-containing dipeptidyl carboxypeptidase widely distributed in mammalian tissues and is thought to play a critical role in blood pressure regulation. The predicted protein is identical, from residue 37 to its C terminus, to the second half or C-terminal domain of the endothelial ACE sequence. The protein sequence inferred consists of a 732-residue preprotein including a 31-residue signal peptide. The mature polypeptide has a molecular weight of 80,073. Although ACE has been studied primarily in the context of its role in blood pressure regulation, this widely distributed enzyme has many other physiological functions. The ACE gene encodes two isozymes. The somatic isozyme is expressed in many tissues, including vascular endothelial cells, renal epithelial cells, and testicular Leydig cells, whereas the testicular or germinal angiotensin-converting enzyme is expressed only in sperm. The standard product used in this kit is recombinant human ACE, consisting of 30-1261 amino acids with the molecular mass of 120KDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 313pg/ml, 156pg/ml human ACE standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernatants, serum, plasma(heparin) or saliva to each empty well. We recommend that each human ACE standard solution and each sample is measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty