Would you like to log in again?
Mouse Amphiregulin - AR ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0361
76236-868EA 636 USD InStock
76236-868
Mouse Amphiregulin - AR ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse Amphiregulin - AR AccuSignal ELISA Kit
  • Natural and recombinant mouse Amphiregulin.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: V100-A248
Amphiregulin, also known as AREG, is a protein that is encoded by the AREG gene. The protein encoded by this gene is a member of the epidermal growth factor family. This gene is mapped to mouse chromosome 5. It is an autocrine growth factor as well as a mitogen forastrocytes, Schwann cells, fibroblasts. Amphiregulin is related to toepidermal growth factor(EGF) and transforming growth factor alpha(TGF-alpha). This protein interacts with the epidermal growth factor receptor to promote the growth of normal epithelial cells. It has been found that transgene integration and subsequent expression of Amphiregulin in basal keratinocytes correlated with a psoriasis-like skin phenotype, and its expression was increased in parallel with typical Th2 cytokines. Amphiregulin is also an important paracrine mediator of estrogen function specifically required for puberty-induced ductal elongation but not for any earlier or later developmental stages, and it can enhance resistance to nematodes.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.3pg/ml, 15.6pg/ml mouse Amphiregulin standard solutions into the pre-coated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. We recommend that each mouse Amphiregulin standard solution and each sample is measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty