Would you like to log in again?
Human CCL18 - PARC ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0396
76236-882EA 636 USD InStock
76236-882
Human CCL18 - PARC ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human CCL18 - PARC AccuSignal ELISA Kit
  • Natural and recombinant human CCL18.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: A21-A89
Chemokine(C-C motif) ligand 18(CCL18) is a small cytokine belonging to the CC chemokine family that was previously called PARC(pulmonary and activation-regulated chemokine). CCL18 is approximately 60% identical in amino acid sequence to CCL3. By analysis of a previously mapped CCL18 from 17q11.2, it is determined that the PARC gene is located within 1 of the 2 clusters of CC chemokine genes in this region. It is expressed at high levels in lung and at lower levels in certain lymphoid tissues, such as the lymph nodes, and is chemotactic for activated T cells and nonactivated lymphocytes. Beside, CCL18 recruits Th2 cells and basophils and may play a predominant role in allergic asthma.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.3pg/ml, 15.6pg/ml, 7.8pg/ml human CCL18 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates or serum to each empty well. We recommend that each human CCL18 standard solution and each sample is measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty