Would you like to log in again?
Mouse CSF1R - M-CSFR ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0479
76236-902EA 636 USD InStock
76236-902
Mouse CSF1R - M-CSFR ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse CSF1R - M-CSFR AccuSignal ELISA Kit
  • Natural and recombinant mouse CSF1R.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: A20-S511
CSF1R(Colony stimulating factor 1 receptor),also known as M-CSFR and CD115, is a cell-surface protein encoded, in humans, by the CSF1R gene. The gene is located on long arm of chromosome 5(5q32) on the Crick(minus) strand. The encoded protein is a tyrosine kinase transmembrane receptor and member of the CSF1/PDGF receptor family of tyrosine-protein kinases. The encoded protein is a single pass type I membrane protein and acts as the receptor for colony stimulating factor 1, a cytokine which controls the production, differentiation, and function of macrophages. Both CSF1R, and its ligand colony stimulating factor 1 play an important role in the development of the mammary gland and may be involved in the process of mammary gland carcinogenesis.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml mouse CSF1R standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse CSF1R standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty