Would you like to log in again?
Human Mer ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0481
76236-906EA 724 USD InStock
76236-906
Human Mer ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Mer AccuSignal ELISA Kit
  • Natural and recombinant human Mer.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: sf21; Immunogen sequence: R26-A499
Proto-oncogene tyrosine-protein kinase MER is an enzyme that in humans is encoded by the MERTK gene. This gene is a member of the MER/AXL/TYRO3 receptor kinase family and encodes a transmembrane protein with two fibronectin type-III domains, two Ig-like C2-type(immunoglobulin-like) domains, and one tyrosine kinase domain. Its gene is mapped to chromosome 2q14.1. Mer encodes a 984-amino acid protein with a calculated molecular mass of 109 kD. It is expressed in numerous neoplastic B- and T-cell lines. Mutations in this gene have been associated with disruption of the retinal pigment epithelium(RPE) phagocytosis pathway and onset of autosomal recessive retinitis pigmentosa(RP)
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human Mer standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, cell lysates, tissue lysates or serum to each empty well. It is recommended that each human Mer standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty