Would you like to log in again?
Mouse MER - MERTK ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0482
76236-908EA 687 USD InStock
76236-908
Mouse MER - MERTK ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse MER - MERTK AccuSignal ELISA Kit
  • Natural and recombinant mouse MER.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: sf21; Immunogen sequence: E23-F498
Proto-oncogene tyrosine-protein kinase MER, also called MERTK is an enzyme that in humans is encoded by the MERTK gene. This gene is a member of the MER/AXL/TYRO3 receptor kinase family and encodes a transmembrane protein with two fibronectin type-III domains, two Ig-like C2-type(immunoglobulin-like) domains, and one tyrosine kinase domain. This gene is mapped to 2q13. MERTK signaling plays a role in various processes such as macrophage clearance of apoptotic cells, platelet aggregation, cytoskeleton reorganization and engulfment. Functions in the retinal pigment epithelium(RPE) as a regulator of rod outer segments fragments phagocytosis. This gene also plays an important role in inhibition of Toll-like receptors(TLRs)-mediated innate immune response by activating STAT1, which selectively induces production of suppressors of cytokine signaling SOCS1 and SOCS3.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 313pg/ml, 156pg/ml, 78pg/ml mouse MER standard solutions into the pre-coated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, cell lysates, tissue homogenates or serum to each empty well. We recommend that each mouse MER standard solution and each sample is measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty