Would you like to log in again?
Human Lipocalin-2 - NGAL ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0514
76236-916EA 687 USD InStock
76236-916
Human Lipocalin-2 - NGAL ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Lipocalin-2 - NGAL AccuSignal ELISA Kit
  • Natural and recombinant human NGAL.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: Q21-G198
Lipocalin-2(LCN2), also known as NGAL, is a protein associated with neutrophil gelatinase.1 The LCN2 gene is located at 9q34 and contains 7 exons.2 The 25-kD LCN2 protein is believed to bind small lipophilic substances such as bacteria-derived lipopolysaccharide(LPS) and formylpeptides and may function as a modulator of inflammation. NGAL tightly binds bacterial catecholate-type ferric siderophores through a cyclically permuted, hybrid electrostatic/cation-pi interaction and is a potent bacteriostatic agent in iron-limiting conditions.3 The primary LCN2 transcript is 3,696 nucleotides long, and the processed transcript is 809 nucleotides long.4 LCN2 expression in adult bone marrow, uterus, prostate, salivary gland, stomach, appendix, colon, trachea, and lung, and in fetal spleen and lung.The standard product used in this kit is recombinant human NGAL, consisting of 178 amino acids with the molecular mass of 22KDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human NGAL standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma(heparin), saliva or urine to each empty well. It is recommended that each human NGAL standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty