Would you like to log in again?
Mouse TNFSF4 - OX40L ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0519
76236-926EA 636 USD InStock
76236-926
Mouse TNFSF4 - OX40L ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse TNFSF4 - OX40L AccuSignal ELISA Kit
  • Natural and recombinant mouse TNFSF4.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: S51-L198
OX40L is the ligand for CD134 and is expressed on such cells as DC2s enabling amplification of Th2 cell differentiation. OX40L has also been designated CD252. By analysis of an interspecific backcross, Baum et al.(1994) mapped the mouse Tnfsf4 gene to chromosome 1. Using fluorescence in situ hybridization, they localized the human TNFSF4 gene to 1q25, a region sharing homology of synteny with the portion of mouse chromosome 1 containing the Tnfsf4 gene.OX40L strongly inhibited the generation of IL-10-producing Tr1 cells induced by two physiologic stimuli, the inducible costimulatory ligand and immature dendritic cells. In addition, OX40L strongly inhibited IL-10 production and suppressive function of differentiated IL-10-producing Tr1 cells. Tnfsf4 underlies Ath1 in mice and that polymorphisms in its human homolog TNFSF4 increase the risk of myocardial infarction in humans.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml mouse TNFSF4 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse TNFSF4 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty