Would you like to log in again?
Human PTX3 - Pentraxin 3 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0521
76236-930EA 636 USD InStock
76236-930
Human PTX3 - Pentraxin 3 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human PTX3 - Pentraxin 3 AccuSignal ELISA Kit
  • Natural and recombinant human PTX3.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: E18-S381
PTX3(Pentraxin 3) is a member of the pentraxin superfamily. This super family characterized by cyclic multimeric structure. The PTX3 gene is mapped to 3q25.32. The predicted 381-amino acid PTX3 protein has homology to the pentraxin protein family. Significant levels of PTX3 were detected in plasma of neutropenic patients with systemic A. PTX3 is effective in preventing CMV infection and reactivation, as well as subsequent Aspergillus infection. PTX3 activates the classical pathway of complementactivation and facilitates pathogen recognition by macrophages and DCs.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 20000pg/ml, 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml human PTX3 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma(EDTA) or saliva to each empty well. It is recommended that each human PTX3 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty