Would you like to log in again?
Mouse TNFRSF9 - 4-1BB ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0655
76236-940EA 636 USD InStock
76236-940
Mouse TNFRSF9 - 4-1BB ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse TNFRSF9 - 4-1BB AccuSignal ELISA Kit
  • Natural and recombinant mouse TNFRSF9.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: V24-L187
CD137 is a member of the tumor necrosis factor(TNF) receptor family. Its alternative names are tumor necrosis factor receptor superfamily member 9(TNFRSF9), 4-1BB and induced by lymphocyte activation(ILA). CD137 gene was localized to chromosome 1p36.Members of this receptor family and their structurally related ligands are important regulators of a wide variety of physiological processes and play an especially important role in the regulation of immune responses. CD137 can be expressed by activated T cells, but to a larger extent on CD8 than on CD4 T cells. The best characterized activity of CD137 is its costimulatory activity for activated T cells. Crosslinking of CD137 enhances T cell proliferation, IL-2 secretion survival and cytolytic activity. Further, it can enhance immune activity to eliminate tumors in mice.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml mouse TNFRSF9 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse TNFRSF9 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty