Would you like to log in again?
Mouse CD163 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0658
76236-944EA 636 USD InStock
76236-944
Mouse CD163 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse CD163 AccuSignal ELISA Kit
  • Natural and recombinant mouse CD163.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: V39-T1045
CD163(Cluster of Differentiation 163) is a human protein encoded by the CD163 gene. It has also been shown to mark cells of monocyte/macrophage lineage. CD163, a member of the scavenger receptor cysteine-rich(SRCR) superfamily, is exclusively expressed by monocytes and macrophages. Using FISH, somatic cell hybrid analysis, and radiation hybrid analysis, CD163 gene was mapped the to chromosome 12p13.3. CD163 is upregulated in a large range of diseases inflammatory diseases including type 2 diabetes, macrophage activation sickness, Tangier's disease, reumatoid arthritis etc.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 50ng/ml, 25ng/ml, 12.5ng/ml, 6.25ng/ml, 3.12ng/ml, 1.56ng/ml, 0.78ng/ml mouse CD163 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum, plasma(heparin) to each empty well. We recommend that each mouse CD163 standard solution and each sample is measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty