Would you like to log in again?
Mouse S100A8 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0661
76236-950EA 636 USD InStock
76236-950
Mouse S100A8 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse S100A8 AccuSignal ELISA Kit
  • Natural and recombinant mouse S100A8.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: P2-E89
S100 calcium-binding protein A8 (S100A8), also known as calgranulin A, is a protein that in humans is encoded by the S100A8 gene. It is mapped to chromosome 1q21.3 based on an alignment of the S100A8 sequence with the genomic sequence. The proteins S100A8 and S100A9 form a heterodimer called calprotectin which is a major calcium- and zinc-binding protein in the cytosol of neutrophils, monocytes, and keratinocytes. This gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. And S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in the inhibition of casein kinase and as a cytokine. Altered expression of this protein is associated with the disease cystic fibrosis.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml mouse S100A8 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse S100A8 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty