Would you like to log in again?
Mouse S100A9 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0662
76236-952EA 636 USD InStock
76236-952
Mouse S100A9 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse S100A9 AccuSignal ELISA Kit
  • Natural and recombinant mouse S100A9.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: A2-K113
Protein S100A9 also known as migration inhibitory factor-related protein 14 (MRP-14) or calgranulin-B is a protein that is encoded by the S100A9 gene. S100A9 is a member of the S100 family of proteins containing 2 EF hand calcium-binding motifs. It is mapped to Chromosome 3 in mouse. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. This protein may function in the inhibition of casein kinase. S100A9 complexes with S100A8, another member of the S100 family of calcium-modulated proteins regulates myeloid cell function by binding to Toll-like receptor-4 (TLR-4) and the receptor for advanced glycation end products.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml mouse S100A9 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse S100A9 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty