Would you like to log in again?
Human Azurocidin ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0667
76236-962EA 636 USD InStock
76236-962
Human Azurocidin ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Azurocidin AccuSignal ELISA Kit
  • Natural and recombinant human Azurocidin.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: I27-P248
Azurocidin, also known as cationic antimicrobial protein CAP37 or heparin-binding protein (HBP), is a protein that in humans is encoded by the AZU1 gene. This encoded protein is a member of the serine protease gene family, but it is not a serine proteinase, because the active site serine and histidine residues are replaced. Azurocidin is mapped to 19p13.3. The protein encoded by this gene is an azurophil granule antibiotic protein, with antibacterial activity. It is also an important multifunctional inflammatory mediator. In addition to it, Azurocidin is also a specific chemoattractant for monocytes. It lacks the chemotactic activity for neutrophils and lymphocytes, and this gene is probably responsible for the wave of monocytes that follows the initial wave of PMNs typical of the inflammatory response.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml human Azurocidin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human Azurocidin standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty