Would you like to log in again?
Human Ficolin-2 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0742
76236-976EA 636 USD InStock
76236-976
Human Ficolin-2 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Ficolin-2 AccuSignal ELISA Kit
  • Natural and recombinant human Ficolin-2.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: L26-A313
Ficolin-2, which was initially identified as L-ficolin, is a protein that in humans is encoded by the FCN2 gene. The product of this gene belongs to the ficolin family of proteins. This family is characterized by the presence of a leader peptide, a short N-terminal segment, followed by a collagen-like region, and a C-terminal fibrinogen-like domain. It is mapped to 9q34.3. This gene is predominantly expressed in the liver, and has been shown to have carbohydrate binding and opsonic activities. Serum concentration of ficolin-2 varies considerably in healthy individuals. FCN2 promoter polymorphisms were associated with marked changes in theficolin-2 serum concentration.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 50ng/ml, 25ng/ml, 12.5ng/ml, 6.25ng/ml, 3.12ng/ml, 1.56ng/ml, 0.78ng/ml human Ficolin-2 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin) to each empty well. It is recommended that each human Ficolin-2 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty