Would you like to log in again?
Mouse VE-Cadherin - CD144 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0791
76236-986EA 636 USD InStock
76236-986
Mouse VE-Cadherin - CD144 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse VE-Cadherin - CD144 AccuSignal ELISA Kit
  • Natural and recombinant mouse VE-Cadherin.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: D46-Q599
Cadherin 5, type 2 or VE-cadherin also known as CD144, is encoded by the gene CDH5. It is a classical cadherin from the cadherin superfamily. This gene is mapped to mouse chromosome 8. The encoded protein is a calcium-dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Functioning as a classic cadherin by imparting to cells the ability to adhere in a homophilic manner, the protein may play an important role in endothelial cell biology through control of the cohesion and organization of the intercellular junctions. It has found that PECAM1, VE-cadherin and VEGFR2 together are sufficient to confer responsiveness to flow in heterologous cells.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 20,000pg/ml, 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml mouse VE-Cadherin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse VE-Cadherin standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty