Would you like to log in again?
Human DKK-3 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0794
76236-992EA 636 USD InStock
76236-992
Human DKK-3 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human DKK-3 AccuSignal ELISA Kit
  • Natural and recombinant human DKK-3.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: A22-I350
Dickkopf-related protein 3 is a protein that in humans is encoded by the DKK3 gene. This gene encodes a protein that is a member of the dickkopf family. It is mapped to 11p15.3. The secreted protein contains two cysteine rich regions and is involved in embryonic development through its interactions with the Wnt signaling pathway. The expression of this gene is decreased in a variety of cancer cell lines and it may function as a tumor suppressor gene. Members of the Dkk-related family display unique patterns of mRNA expression in human and mouse tissues, and are secreted when expressed in 293T cells. DKKs play an important role in vertebrate development, where they locally inhibit Wnt regulated processes such as antero-posterior axial patterning, limb development, somitogenesis and eye formation. In the adult, Dkks are implicated in bone formation and bone disease, cancer and Alzheimer disease.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human DKK-3 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human DKK-3 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty