Would you like to log in again?
HyClone™ HyCell CHO Cell Culture Media, HyClone products (Cytiva)
Supplier: HyClone products (Cytiva)
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
HyClone™
SH30933.02 SH30933.02 SH30934.01 SH30933.01 SH30934.01 SH30933.01
76237-084EA 119.26 USD InStock
CA76237-084 CA76237-082 76237-084 76237-082 CA76237-080 76237-080
HyClone™ HyCell CHO Cell Culture Media, HyClone products (Cytiva)
Cell Culture Media
HyCell™ CHO media has been developed for CHO cell culture for bioprocess applications.
  • Animal-derived component free (ADCF)
  • Allows for direct or sequential adaptation
  • Component traceability
  • Manufactured under cGMP
This media has been specifically formulated using Metabolic Pathway Design. It provides optimal performance and supports higher peak cell density, enhanced product yield, and quality. The versatility of HyCell™ CHO media allows quick adaptation and supports exceptional growth, high density, and high productivity up to five-fold or more over other comparable media across a broad variety of CHO clones.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty