Would you like to log in again?
QuantiChrom™ Arginase Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
DARG-100
76237-100EA 409.2 USD InStock
76237-100
QuantiChrom™ Arginase Assay Kit, BioAssay Systems
Assays Enzyme Assays
For quantitative determination of arginase activity and screen for its inhibitors.
  • Sensitive and accurate
  • Detection limit: 0.3 U/ for 2 hr arginase reaction in 96-well assay format
  • Simple and high-throughput
  • Can be readily automated as a high-throughput assay for thousands of samples per day
  • Method: OD430 nm
  • Samples: Enzyme preparations, serum, plasma, tissue culture and more
Arginase (L-arginine ureohydrolase EC 3.5.3.1) is present in mammals and plants. In humans, arginase is expressed predominantly in the liver, and to lesser degrees in breast, kidney, testes, salivary glands, epidermis and erythrocytes. Arginase catalyzes the conversion of arginine to ornithine and urea, completing the last step in the urea cycle. Arginase activity is a key diagnostic indicator. Increased levels of arginase activity in blood have been associated with liver damage.
The procedure involves incubation of the provided substrate with the sample in a microplate followed by the addition of the coloring reagent. Hyperargininemia due to arginase deficiency is an inherited autosomal recessive disease. Simple, direct and automation-ready procedures for measuring arginase activity in biological samples are highly desirable in Research and Drug Discovery. Arginase assay kit provides a sensitive and convenient method for arginase activity determination. The method utilizes a chromogen that forms a colored complex specifically with urea produced in the arginase reaction. The intensity of the color is directly proportional to the arginase activity in the sample.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty