Would you like to log in again?
QuantiChrom™ α-L-Fucosidase Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
DFUC-100
76237-104EA 590.41 USD InStock
76237-104
QuantiChrom™ α-L-Fucosidase Assay Kit, BioAssay Systems
Assays Enzyme Assays
For quantitative determination of α-L-Fucosidase activity in biological samples (example: plasma, serum, tissue, cell lysate, and more.)
  • High sensitivity and wide linear range, linear detection range (10 μl sample): 1 to 100 U/l
  • Homogeneous and simple procedure
  • simple add-mix-read procedure allows reliable quantitation of fucosidase activity within 20 minutes
  • Robust and amenable to HTS
  • Method:OD405 nm
  • Samples: Plasma, serum, tissue, cell lysate, and more
All reagents are compatible with high-throughput liquid handling instruments. Can be readily automated to measure thousands of samples per day. α-L-Fucosidase (AFU) is an enzyme coded by the FUCA1 gene in humans and catalyzes the breakdown of L-Fucose. A genetic deficiency in this enzyme results in a neurovisceral storage disease, fucosidosis, which is characterized by the accumulation of fucose. Low serum activity of fucosidase has also been linked to ovarian caricinoma. Elevated fucosidase serum activity has been observed in patients with diabetes, hyperthyroidism, cirrhosis, and hepatitis. Increased activity has been associated with lung, breast, stomach, ovary, uterus, and liver carcinomas.
Non-radioactive, colorimetric AFU assay is based on the cleavage of 4-nitrophenol from the synthetic substrate. Nitrophenol becomes intensely colored after addition of the stop reagent. The increase in absorbance at 405 nm after addition of the stop reagent is directly proportional to the enzyme activity.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty