Would you like to log in again?
Mouse BMP-2 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0152
76237-116EA 687 USD InStock
76237-116
Mouse BMP-2 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse BMP-2 AccuSignal ELISA Kit
  • Natural and recombinant mouse BMP-2.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard:CHO ; Immunogen sequence: Q283-R398
Bone morphogenetic protein-2(BMP-2, BMP 2A) belongs to the transforming growth factor beta(TGF-beta) superfamily. It is thought to be involved in cartilage and bone formation during embryogenesis, but may have additional functions in morphogenesis as implied by its expression in various organs and embryonic tissues of mice. BMP-2 has been identified as a candidate mediator of retinoid activity. BMP-2 protein induces medulloblastoma cell apoptosis, whereas the BMP-2 antagonist noggin blocks both retinoid and BMP-2-induced apoptosis. BMP-2 also induces p38 mitogen-activated protein kinase(MAPK), which is necessary for BMP-2 and retinoid-induced apoptosis. Bone morphogenic proteins(BMPs) are known to promote osteogenesis, and clinical trials are currently underway to evaluate the ability of certain BMPs to promote fracture-healing and spinal fusion. The standard product used in this kit is recombinant mouse BMP-2, constituting dimer by two chains of 114 amino acids with the molecular mass of 26KDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml mouse BMP-2 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse bone tissue, cell culture supernates or serum to each empty well. It is recommended that each mouse BMP-2 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty