Would you like to log in again?
Human EGF ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0156
76237-124EA 636 USD InStock
76237-124
Human EGF ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human EGF AccuSignal ELISA Kit
  • Natural and recombinant human EGF.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: N971-R1023
Epidermal growth factor or EGF is a growth factor that plays an important role in the regulation of cell growth, proliferation, and differentiation by binding to its receptor EGFR. Human EGF is a 6045-Da protein with 53 amino acid residues and three intramolecular disulfide bonds. EGF acts by binding with high affinity to epidermal growth factor receptor(EGFR) on the cell surface and stimulating the intrinsic protein-tyrosine kinase activity of the receptor. EGF results in cellular proliferation, differentiation, and survival. It also has a profound effect on the differentiation of specific cells in vivo and is a potent mitogenic factor for a variety of cultured cells of both ectodermal and mesodermal origin. EGF has strong expression in kidney, salivary gland, cerebrum, and prostate, moderate expression in trachea and thyroid, and low expression in bone marrow, heart, spleen, thymus, uterus, and colon. No expression was detected in adrenal gland, liver, lung, cerebellum, placenta, and small intestine.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 300pg/ml, 150pg/ml, 75pg/ml, 37.5pg/ml, 18.8pg/ml, 9.4pg/ml, 4.7pg/ml human EGF standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma( heparin, EDTA, citrate) or urine to each empty well. It is recommended that each human EGF standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty